Order Entry
Puerto Rico
ContactUsLinkComponent
Guanidinium hydrochloride 99.5%, without anti-caking agent
Irritant
Guanidinium hydrochloride 99.5%, without anti-caking agent
Certificates
undefined
Irritant
Guanidinium hydrochloride 99.5%, without anti-caking agent

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specification Test Results

  • Appearance:
    Colorless to white Crystals
  • Infrared spectrum:
    Conforms to structure and standard
  • Melting point:
    178 to 188 °C (Identification C)
  • UV:
    (6 M in water)
  • Identification A:
    Passes test
  • Identification B:
    Passes test
  • Titration with HClO4:
    ≥99.5 %

Specification Test Results

Frequently Bought Together