Sodium acetate trihydrate 99.5% for analysis
Supplier: Thermo Scientific Chemicals
Certificates
About this item
CAS Number:6131-90-4
:
Formula:H₃CCOONa·3H₂O
MDL Number:MFCD00071557
:136.08 g/mol
Merck Index:13,08642
Boiling Pt:120…123 °C (1013 hPa)
Melting Pt:58 °C
Storage Temperature:Ambient
EINECS:204-823-8
Specification Test Results
- Appearance:White Crystals
- Infrared spectrum:Conforms
- pH:7.5 to 9.0 (5 % at 20°C)
- Titration with HClO4:≥99.4 %
- Chloride (Cl):≤10 ppm
- Phosphate (PO4):≤5 ppm
- Sulfate (SO4):≤20 ppm
- Aluminium (Al):≤5 ppm
- Calcium (Ca):≤20 ppm
- Iron (Fe):≤5 ppm
- Magnesium (Mg):≤20 ppm
- Potassium (K):≤50 ppm
- Heavy metals (as Pb):≤5 ppm
- Insoluble matter:≤0.005 % (in water)
- KMnO4-reducing substance:≤0.005 % (as HCOOH)