Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
L(+)-Ascorbic acid 99% ACS

L(+)-Ascorbic acid 99% ACS

Irritant
Health
Flammable
L(+)-Ascorbic acid 99% ACS
Certificates
L(+)-Ascorbic acid 99% ACS

3 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
  • CAS Number:50-81-7
  • Formula:C₆H₈O₆
  • MDL Number:MFCD00064328
  • Molecular Weight:176.13 g/mol
  • Merck Index:13,00837
  • Boiling Pt:62 °C (1013 hPa)
  • Melting Pt:–136 °C (dec.)
  • Density:0.68 g/cm³
  • Flash Pt:–26 °C
  • Storage Temperature:Refrigerator
  • EINECS:212-108-7
  • UN:3295
  • ADR:3,II
  • REACH:exempt
Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3910 to 1.3930 (20°C, 589 nm)
  • GC:
    ≥98.5 %
  • Specific optical rotation:
    +20.5° to +21.5° (25°C, 589 nm) (c=10, H2O)
  • Iron (Fe):
    ≤10 ppm
  • Heavy metals (ICP-OES):
    ≤20 ppm
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Plastic bottle
    100 g
    Log in for availability
    Log in for availability
    Each (100g)
    Each (100g) - $89.50
    $89.50
    Each (100g)
       $89.50

    Supplier Number: 40147-1000
  • Plastic bottle
    500 g
    Log in for availability
    Log in for availability
    Each (500g)
    Each (500g) - $293.00
    $293.00
    Each (500g)
       $293.00

    Supplier Number: 40147-5000
  • Plastic bottle
    2.5 kg
    Log in for availability
    Log in for availability
    Each (2,500g/mol)
    Each (2,500g/mol) - $1,200.00
    $1,200.00
    Each (2,500g/mol)
       $1,200.00

    Supplier Number: 40147-0025

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.