


Tetrahydrofuran 99.6% stabilized ACS
Supplier: Thermo Scientific Chemicals
New Product
Certificates
About this item
CAS Number:109-99-9
:
Formula:C₄H₈O
MDL Number:MFCD00005356
:72.11 g/mol
Merck Index:12,09351
Boiling Pt:66 °C (760 mmHg)
Melting Pt:–108.4 °C
Density:0.88 g/cm³
Flash Pt:–21 °C
UN:2056
ADR:3,II
:Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
Storage Temperature:Ambient
EINECS:203-726-8
REACH:05-2114615188-48
:GHS02
:GHS07
:GHS08
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Appearance:Clear liquid
- Water:≤0.03 % (Coulometric)
- GC:≥99.5 %
- Color scale:≤10 APHA
- Stabilizer:200 to 400 ppm BHT
- Residue after ignition:≤0.002 %
- Acidity/alkalinity:≤0.0002 meq/g
- Calcium (Ca):≤0.5 ppm
- Copper (Cu):≤0.05 ppm
- Iron (Fe):≤0.5 ppm
- Lead (Pb):≤0.05 ppm
- Peroxides:≤0.0025 %