Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Sodium hydroxide, pellets, pure

Sodium hydroxide, pellets, pure

Corrosive
Sodium hydroxide, pellets, pure
Certificates
Sodium hydroxide, pellets, pure

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  Caustic soda
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol
  • Boiling Pt:1390 °C (760 mmHg)
  • Melting Pt:318 °C
  • Storage Temperature:Ambient
  • UN:1823
  • ADR:8,II
Specifications for Product Family

Specification Test Results

  • Appearance:
    White to off-white Pellets
  • Titration with HCl:
    ≥97.5 %
  • Chloride (Cl):
    ≤0.005 %
  • Sulfate (SO4):
    ≤0.005 %
  • Sodium carbonate (Na2CO3):
    ≤2.0 %
  • Iron (Fe):
    ≤0.001 %
  • Heavy metals (as Pb):
    ≤20 ppm
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Plastic bottle
    5 kg
    Log in for availability
    Log in for availability
    Each (5kg)
    Each (5kg) - $90.00
    $90.00
    Each (5kg)
       $90.00
    Minimum order quantity is 1

    Supplier Number: 44728-0050
  • Fibre drum
    25 kg
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $308.00
    $308.00
    Each(1/Itm/Bx)
       $308.00

    Supplier Number: 447280250

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.