Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Potassium bromide 99+% for analysis

Potassium bromide 99+% for analysis

Irritant
Potassium bromide 99+% for analysis
Certificates
Potassium bromide 99+% for analysis

3 Options Available Below

Product Details & Documents
  • CAS Number:7758-02-3
  • Formula:KBr
  • MDL Number:MFCD00011358
  • Molecular Weight:119 g/mol
  • Merck Index:13,07701
  • Boiling Pt:1435 °C (760 mmHg)
  • Melting Pt:734 °C
  • Storage Temperature:Ambient
  • EINECS:231-830-3
Specifications for Product Family

Specification Test Results

  • Appearance:
    White Crystalline powder or crystals and/or chunks
  • Titration Argentometric:
    ≥99.0 %
  • Loss on drying:
    ≤0.5 % (105°C)
  • pH:
    5.0 to 8.8 (5 % at 25°C)
  • Chloride (Cl):
    ≤0.2 %
  • Iodide (I):
    ≤0.05 %
  • Bromate (BrO3):
    ≤0.001 %
  • Sulfate (SO4):
    ≤0.005 %
  • Barium (Ba):
    ≤0.002 %
  • Calcium (Ca):
    ≤0.002 %
  • Iron (Fe):
    ≤5 ppm
  • Sodium (Na):
    ≤0.1 %
  • Heavy metals (as Pb):
    ≤5 ppm
  • Insoluble matter:
    ≤0.005 % (in water)
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Plastic bottle
    100 g
    Log in for availability
    Log in for availability
    Each (100g)
    Each (100g) - $37.40
    $37.40
    Each (100g)
       $37.40
    Minimum order quantity is 1

    Supplier Number: AC22255-1000
  • Plastic bottle
    500 g
    Log in for availability
    Log in for availability
    Each (500g)
    Each (500g) - $101.00
    $101.00
    Each (500g)
       $101.00
    Minimum order quantity is 1

    Supplier Number: 22255-5000
  • Plastic bottle
    2.5 kg
    Log in for availability
    Log in for availability
    Each (2,500g)
    Each (2,500g) - $376.00
    $376.00
    Each (2,500g)
       $376.00
    Minimum order quantity is 1

    Supplier Number: 22255-0025