


p-Xylene 99% for spectroscopy
Catalog # TS42267-0010
Supplier: Thermo Scientific Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:1 L
- Cat. No.:TS42267-0010
Specifications
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- GC:≥98.5 %
- UV:(neat) 1 cm cell vs water
- Color scale:≤15 APHA
Specification Test Results
About this item
:
:106.17 g/mol
Merck Index:12,10214
Boiling Pt:138.3 °C (1013 hPa)
Melting Pt:13 °C
Density:0.866 g/cm³
Flash Pt:25 °C
EINECS:203-396-5
UN:1307
ADR:3,III
:GHS02
:GHS07
:GHS08
Storage Temperature:Ambient
CAS Number:106-42-3
Formula:C₆H₄(CH₃)₂
MDL Number:MFCD00008556