3-Nitrobenzoic acid 99%
Plastic bottle, 5 kg
Catalog # TS12845-0051
Supplier: Thermo Scientific Chemicals
: 12845-0051
3-Nitrobenzoic acid 99%
Catalog # TS12845-0051
Supplier: Thermo Scientific Chemicals
CAS Number: 121-92-6
Jump to:
Product Details & Documents
Synonyms: m-Nitrobenzoic acid
- CAS Number:121-92-6
- Formula:C₇H₅NO₄
- MDL Number:MFCD00007251
- Molecular Weight:167.12 g/mol
- Merck Index:12,06686
- Melting Pt:139…143 °C
- Flash Pt:190 °C
- Storage Temperature:Ambient
- EINECS:204-508-5
Specifications
Specification Test Results
- Appearance:White to light yellow or light green Powder or crystals
- Infrared spectrum:Conforms
- Melting point:139 to 143 °C
- HPLC:≥98.5 %
- Titration with NaOH:98.5 to 101.5 %
- Specific optical rotation:-56° to -61° (20 °C, 589 nm) (c=1, chloroform)
Specifications
- Pack type:Plastic bottle
- Size:5 kg
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



