Order Entry
Puerto Rico
ContactUsLinkComponent
 
 
 
 
1-Butanol 99.5% ACS, meets analytical specifications of Reagent Ph. Eur.
    
New Product
 
undefined
 
 
 
1-Butanol 99.5% ACS, meets analytical specifications of Reagent Ph. Eur.
    
New Product

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental Certification:
    Chem21 solvent selection guide recommended by default and after discussion

 

 

  • Appearance:
    Clear liquid
  • Water:
    ≤0.1 % (K.F.)
  • Boiling point:
    116 to 119 °C
  • Density:
    (20/20 °C) 0.809 to 0.812
  • GC:
    ≥99.4 %
  • Color scale:
    ≤10 APHA
  • Residue after evaporation:
    ≤0.005 %
  • Carbonyl compounds:
    ≤0.01 % (as butyraldehyde)
  • Titrable acid:
    ≤0.0008 meq/g
  • Butyl ether:
    ≤0.2 % (GC)

 

Frequently Bought Together