Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Diethyl ether 99+% stabilised, extra pure

Diethyl ether 99+% stabilised, extra pure

Irritant
Flammable
Diethyl ether 99+% stabilised, extra pure
Certificates
Diethyl ether 99+% stabilised, extra pure

3 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane
  • CAS Number:60-29-7
  • Formula:(CH₃CH₂)₂O
  • MDL Number:MFCD00011646
  • Molecular Weight:74.12 g/mol
  • Boiling Pt:34.6 °C (760 mmHg)
  • Melting Pt:–116 °C
  • Density:0.714 g/cm³
  • Flash Pt:–45 °C
  • Storage Temperature:Ambient
  • UN:1155
  • ADR:3,I
  •  :Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3510 to 1.3540 (20°C, 589 nm)
  • Water:
    ≤0.1 % (Coulometric)
  • GC:
    ≥99.0 %
  • Color scale:
    ≤10 APHA
  • Stabilizer:
    5 to 7 ppm BHT
  • Relative density:
    (20°C) 0.714 to 0.716
  • Residue after evaporation:
    ≤0.005 %
  • Acidity:
    ≤0.001 meq/g
  • Peroxides:
    ≤0.0005 %
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $101.00
    $101.00
    Each(1/Itm/Bx)
       $101.00

    Supplier Number: 176830010
  • Metal drum
    5 L
    Log in for availability
    Log in for availability
    Each (5l)
    Each (5l) - $279.00
    $279.00
    Each (5l)
       $279.00

    Supplier Number: 17683-0050
  • Metal drum
    25 L
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $1,190.00
    $1,190.00
    Each(1/Itm/Bx)
       $1,190.00

    Supplier Number: 176830250

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.