1-Chlorobutane 99.8% for HPLC
Supplier: Thermo Scientific Chemicals
Certificates
About this item
CAS Number:109-69-3
:
Formula:CH₃(CH₂)₃Cl
MDL Number:MFCD00001009
:92.57 g/mol
Boiling Pt:78.4 °C (1013 hPa)
Melting Pt:–123 °C
Density:0.88 g/cm³
Flash Pt:–12 °C
Storage Temperature:Ambient
UN:1127
ADR:3,II
:GHS02
:GHS08
Specification Test Results
- Appearance:Colorless Liquid
- Water:≤0.020 %
- GC:≥99.7 %
- UV:1 cm cell vs HPLC-grade water
- Color scale:≤10 APHA
- Residue after evaporation:≤2 ppm
- Acidity:≤0.0003 meq/g
- Filtered to 0.2 micrometer:Filtered