Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Acetone, Technical Grade

Acetone, Technical Grade

Irritant
Flammable
   Sustainable Options Available
Acetone, Technical Grade
   Sustainable Options Available
Certificates
Acetone, Technical Grade

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  2-Propanone, DMK, Propanone, Dimethyl ketone
  • CAS Number:67-64-1
  • Formula:CH₃COCH₃
  • MDL Number:MFCD00008765
  • Molecular Weight:58.08 g/mol
  • Boiling Pt:56 °C (760 mmHg)
  • Melting Pt:–95 °C
  • Density:0.79 g/cm³
  • Flash Pt:–20 °C
  • Storage Temperature:Ambient
  • UN:1090
  • ADR:3,II
Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Refractive index:
    1.3580 to 1.3600 (20°C, 589 nm)
  • Density:
    0.790 to 0.792 kg/l
  • GC:
    ≥98.5 %

Specifications

Specifications

  • Catalog No:
  • TS44415-0050
  • TS44415-0250
  • Environmentally Preferable:
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Environmental Certification:
  • Chem21 Solvent selection guide recommended after discussion
  • Chem21 Solvent selection guide recommended after discussion

Specifications

Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Plastic drum
    5 L
    Log in for availability
    Log in for availability
    Each (5l)
    Each (5l) - $85.20
    $85.20
    Each (5l)
       $85.20

    Supplier Number: 44415-0050
    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    TS44415-0050
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental Certification
    Chem21 Solvent selection guide recommended after discussion
      Safer Human and Environmental Health
  • Plastic can
    25 L
    Log in for availability
    Log in for availability
    Each (25l)
    Each (25l) - $219.00
    $219.00
    Each (25l)
       $219.00

    Supplier Number: 44415-0250
    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    TS44415-0250
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental Certification
    Chem21 Solvent selection guide recommended after discussion
      Safer Human and Environmental Health

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.