Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Acetone 99.5+% for analysis

Irritant
Flammable
   Sustainable Options Available
Acetone 99.5+% for analysis
   Sustainable Options Available
Certificates
Acetone 99.5+% for analysis

6 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  2-Propanone, DMK, Propanone, Dimethyl ketone
  • CAS Number:67-64-1
  • Formula:CH₃COCH₃
  • MDL Number:MFCD00008765
  • Molecular Weight:58.08 g/mol
  • Merck Index:13,00067
  • Boiling Pt:56 °C (760 mmHg)
  • Melting Pt:–95 °C
  • Density:0.79 g/cm³
  • Flash Pt:–20 °C
  • Storage Temperature:Ambient
  • EINECS:200-662-2
  • UN:1090
  • ADR:3,II
  • REACH:01-2119471330-49
Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear liquid
  • Water:
    ≤0.5 % (Coulometric)
  • GC:
    ≥99.50 %
  • Color scale:
    ≤5 APHA
  • Residue after evaporation:
    ≤0.001 %
  • Acidity/alkalinity:
    ≤0.0005 meq/g
  • Cadmium (Cd):
    ≤0.05 ppm
  • Copper (Cu):
    ≤0.1 ppm
  • Iron (Fe):
    ≤0.5 ppm
  • Lead (Pb):
    ≤0.1 ppm
  • Magnesium (Mg):
    ≤0.1 ppm
  • Zinc (Zn):
    ≤0.05 ppm
  • KMnO4-reducing substance:
    ≤5 ppm

Specifications

Specifications

  • Catalog No:
  • TS176805000
  • TS176800025
  • TS176800026
  • TS176800010
  • TS17680-0050
  • TS17680-0250
  • Environmentally Preferable:
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Environmental Certification:
  • Chem21 solvent selection guide recommended after discussion
  • Chem21 solvent selection guide recommended after discussion

Specifications

Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Glass bottle
    500 ml
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $45.00
    $45.00
    Each(1/Itm/Bx)
       $45.00

    Supplier Number: 176805000
    Specifications:
    More
    Specifications
    Cat. No.
    TS176805000
    Environmentally Preferable
    Environmental Certification
  • Glass bottle
    2500 ml
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $132.00
    $132.00
    Each(1/Itm/Bx)
       $132.00

    Supplier Number: 176800025
    Specifications:
    More
    Specifications
    Cat. No.
    TS176800025
    Environmentally Preferable
    Environmental Certification
  • Plastic bottle
    2500 ml
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $132.00
    $132.00
    Each(1/Itm/Bx)
       $132.00

    Supplier Number: 176800026
    Specifications:
    More
    Specifications
    Cat. No.
    TS176800026
    Environmentally Preferable
    Environmental Certification
  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $70.70
    $70.70
    Each(1/Itm/Bx)
       $70.70

    Supplier Number: 176800010
    Specifications:
    More
    Specifications
    Cat. No.
    TS176800010
    Environmentally Preferable
    Environmental Certification
  • Plastic drum
    5 L
    Log in for availability
    Log in for availability
    Each (5l)
    Each (5l) - $184.00
    $184.00
    Each (5l)
       $184.00

    Supplier Number: 17680-0050
    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    TS17680-0050
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental Certification
    Chem21 solvent selection guide recommended after discussion
      Safer Human and Environmental Health
  • Metal drum
    25 L
    Log in for availability
    Log in for availability
    Each (25l)
    Each (25l) - $401.00
    $401.00
    Each (25l)
       $401.00

    Supplier Number: 17680-0250
    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    TS17680-0250
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental Certification
    Chem21 solvent selection guide recommended after discussion
      Safer Human and Environmental Health

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.