Acetone 99+%, extra pure
Supplier: Thermo Scientific Chemicals
Acetone 99+%, extra pure
Supplier: Thermo Scientific Chemicals
Certificates
About this item
Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone
- CAS Number:67-64-1
- Formula:CH₃COCH₃
- MDL Number:MFCD00008765
- Molecular Weight:58.08 g/mol
- Merck Index:13,00067
- Boiling Pt:56 °C (760 mmHg)
- Melting Pt:–95 °C
- Density:0.79 g/cm³
- Flash Pt:–20 °C
- Storage Temperature:Ambient
- EINECS:200-662-2
- UN:1090
- ADR:3,II
- REACH:01-2119471330-49
5 Options Available Below
Product Details & Documents
Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone
- CAS Number:67-64-1
- Formula:CH₃COCH₃
- MDL Number:MFCD00008765
- Molecular Weight:58.08 g/mol
- Merck Index:13,00067
- Boiling Pt:56 °C (760 mmHg)
- Melting Pt:–95 °C
- Density:0.79 g/cm³
- Flash Pt:–20 °C
- Storage Temperature:Ambient
- EINECS:200-662-2
- UN:1090
- ADR:3,II
- REACH:01-2119471330-49
Specifications for Product Family
Specification Test Results
- Appearance:Clear liquid
- Water:≤0.5 % (K.F.)
- GC:≥99.0 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.005 %
Specifications
Specifications
- Catalog No:
- TS177170025
- TS177170010
- TS177170050
- TS177170100
- TS17717-0250
- Environmentally Preferable:
- Safer Human and Environmental Health
- Environmental Certification:
- Chem21 Solvent selection guide recommended after discussion
Specifications
Product Family Options
Product Information
- Pack typeSizeAvailabilityPrice
- Supplier Number: 177170025Specifications:
More
Specifications
Cat. No.TS177170025Environmentally PreferableEnvironmental Certification - Supplier Number: 177170010Specifications:
More
Specifications
Cat. No.TS177170010Environmentally PreferableEnvironmental Certification - Supplier Number: 177170050Specifications:
More
Specifications
Cat. No.TS177170050Environmentally PreferableEnvironmental Certification - Supplier Number: 177170100Specifications:
More
Specifications
Cat. No.TS177170100Environmentally PreferableEnvironmental Certification - Supplier Number: 17717-0250Specifications:
Safer Human and Environmental Health More
Specifications
Cat. No.TS17717-0250Environmentally PreferableSafer Human and Environmental HealthEnvironmental CertificationChem21 Solvent selection guide recommended after discussionSafer Human and Environmental Health
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



