
Tetrasodium diphosphate decahydrate 99+% for analysis
Supplier: Thermo Scientific Chemicals
Certificates
About this item
CAS Number:13472-36-1
:
Formula:Na₄P₂O₇·10H₂O
MDL Number:MFCD00149200
:446.06 g/mol
Boiling Pt:93.8 °C (760 mmHg)
Melting Pt:80 °C
Storage Temperature:Ambient
EINECS:231-767-1
:GHS07
Specification Test Results
- Appearance:White Crystals
- Titration with HClO4:≥99.0 %
- Chloride (Cl):≤10 ppm
- Iron (Fe):≤5 ppm
- Potassium (K):≤50 ppm
- Heavy metals (as Pb):≤5 ppm
- Total nitrogen compounds:≤0.001 % (as N)
- Impurity:≤0.1 % (ortho-phosphate (as PO4))
Specification Test Results
Frequently Bought Together