


Toluene-[D₈] 99+ atom % D for NMR spectroscopy
Supplier: Thermo Scientific Chemicals
Certificates
About this item
CAS Number:2037-26-5
:
Formula:C₆D₅CD₃
MDL Number:MFCD00044638
:100.08 g/mol
Boiling Pt:110 °C (1013 hPa)
Melting Pt:–84 °C
Density:0.94 g/cm³ (20 °C)
Flash Pt:4 °C (closed cup)
Storage Temperature:Refrigerator
EINECS:218-009-5
UN:1294
ADR:3,II
:GHS02
:GHS07
:GHS08
Specification Test Results
- Appearance:Clear colorless Liquid
- Water:≤0.05 %
- D-Enrichment:≥99.0 %