Order Entry
United States
ContactUsLinkComponent
Methanol-[D₄] 99.8 atom % D + 0.03% (v/v) TMS for NMR spectroscopy
Methanol-[D₄] 99.8 atom % D + 0.03% (v/v) TMS for NMR spectroscopy
Catalog # TS35147-0075
CAS Number:  
undefined
Methanol-[D₄] 99.8 atom % D + 0.03% (v/v) TMS for NMR spectroscopy
Catalog # TS35147-0075
Supplier Number:  35147-0075
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pack type:
    Ampoule
  • Size:
    10 x 0.75 ml
  • Environmentally Preferable:
  • Cat. No.:
    TS35147-0075

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.05 % (Coulometric)
  • D-Enrichment:
    ≥99.75 %
  • TMS:
    0.020 to 0.040 %

Specification Test Results

About this item

Synonyms: Methanol-[²H₄], Hydroxy methane-D4, Methanol-d4, Methyl-D3 alcohol-D, Carbinol-D4, Wood alcohol-D4, Methyl alcohol-D4, Tetradeuteromethanol
 :
 :36.01 g/mol
Boiling Pt:65.3 °C (1013 hPa)
Melting Pt:–99 °C
Density:0.888 g/cm³ (20 °C)
Flash Pt:12 °C
Storage Temperature:Ambient
EINECS:212-378-6
UN:1230
ADR:3,II
 :GHS06
 :GHS08
 :GHS02
CAS Number:811-98-3
Formula:D₃COD
MDL Number:MFCD00044637