Order Entry
Puerto Rico
ContactUsLinkComponent
Dichloromethane-[D₂] 99.5 atom % D, AcroSeal™ for NMR spectroscopy
Irritant
Health
Dichloromethane-[D₂] 99.5 atom % D, AcroSeal™ for NMR spectroscopy
Catalog # TS43399-1000
CAS Number:  
undefined
Irritant
Health
Dichloromethane-[D₂] 99.5 atom % D, AcroSeal™ for NMR spectroscopy
Catalog # TS43399-1000
Supplier Number:  43399-1000
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, AcroSeal®
  • Size:
    100 ml
  • Cat. No.:
    TS43399-1000

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤50 ppm
  • D-Enrichment:
    ≥99.4 %

Specification Test Results

About this item

 :
 :86.92 g/mol
Boiling Pt:40 °C (760 mmHg)
Melting Pt:–97 °C
Density:1.36 g/cm³ (20 °C)
UN:1593
ADR:6.1,III
 :GHS07
 :GHS08
Storage Temperature:Ambient
CAS Number:1665-00-5
Formula:CD₂Cl₂
MDL Number:MFCD00000882