ContactUsLinkComponent
Acetone-[D₆] 99.8 atom % D for NMR spectroscopy
Irritant
Flammable
Acetone-[D₆] 99.8 atom % D for NMR spectroscopy
Certificates
undefined
Irritant
Flammable
Acetone-[D₆] 99.8 atom % D for NMR spectroscopy
Specification Test Results
  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.03 % (Coulometric)
  • D-Enrichment:
    ≥99.75 %
Specification Test Results