Acetone-[D₆] 99+ atom % D for NMR spectroscopy
:666-52-4
:
:D₃CC(O)CD₃
:MFCD00044635
:64.03 g/mol
:55.5 °C (1013 hPa)
:–93 °C
:0.87 g/cm³
:–20 °C (closed cup)
:Ambient
:211-563-9
:1090
:3,II
:GHS07
:GHS02
- Appearance:Clear colorless Liquid
- Water:≤0.1 % (Coulometric)
- D-Enrichment:≥99.0 %
- HPLC:≥97.5 %