


tert-Butylmagnesium chloride 1.7 M in diethyl ether, AcroSeal™
Catalog # TS39430-8000
Supplier: Thermo Scientific Chemicals
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle, AcroSeal®
- Size:800 ml
- Cat. No.:TS39430-8000
Specifications
Specification Test Results
- Appearance:Dark brown Solution
- Titration Active base:24 to 26 %
Specification Test Results
About this item
:
:116.87 g/mol
Boiling Pt:34 °C (760 mmHg)
Density:0.8 g/cm³
Flash Pt:–40 °C
Storage Temperature:Ambient
UN:3399
ADR:4.3,I
:GHS07
:GHS05
:GHS02
CAS Number:677-22-5
Formula:C₄H₉ClMg
MDL Number:MFCD00000465