



Cyclopropylmagnesium bromide 0.5 M in THF, AcroSeal™
Catalog # TS39761-1000
Supplier: Thermo Scientific Chemicals
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle, AcroSeal®
- Size:100 ml
- Cat. No.:TS39761-1000
Specifications
Specification Test Results
- Appearance:Colorless to light yellow to dark yellow Solution
- Titration Grignard:0.4 to 0.6 M
Specification Test Results
About this item
:
:145.28 g/mol
Flash Pt:–17 °C
Storage Temperature:Ambient
UN:3399
ADR:4.3,I
:GHS07
:GHS05
:GHS02
:GHS08
CAS Number:23719-80-4
Formula:C₃H₅BrMg
MDL Number:MFCD00672094