
Dipotassium tetrachloropalladate > 32.0% Pd
Supplier: Thermo Scientific Chemicals
Certificates
About this item
CAS Number:10025-98-6
:
Formula:Cl₄K₂Pd
MDL Number:MFCD00011373
:326.43
Melting Pt:105 °C
Density:2.67 g/cm³
EINECS:233-049-3
:GHS07
Specification Test Results
- Appearance:Green-brown to dark brown Crystalline powder or crystals
- Assay:≥32.0 % (Pd)
- Calcium (Ca):≤10 ppm
- Copper (Cu):≤10 ppm
- Gold (Au):≤20 ppm
- Iridium (Ir):≤20 ppm
- Iron (Fe):≤10 ppm
- Magnesium (Mg):≤10 ppm
- Platinum (Pt):≤20 ppm
- Rhodium (Rh):≤20 ppm
- Ruthenium (Ru):≤20 ppm
- Silicon (Si):≤30 ppm
- Silver (Ag):≤20 ppm
Specification Test Results
Frequently Bought Together