Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Lead(II) acetate basic ACS

Lead(II) acetate basic ACS

Irritant
Health
Environment
Lead(II) acetate basic ACS
Certificates
Lead(II) acetate basic ACS

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  Horne′s compound, Lead(II) hydroxide acetate, Acetic acid lead (II) salt basic, Lead subacetate
  • CAS Number:1335-32-6
  • Formula:(CH₃COO)₂Pb·2Pb(OH)₂
  • MDL Number:MFCD00011154
  • Molecular Weight:807.72 g/mol
  • Storage Temperature:Ambient
  • UN:2291
  • ADR:6.1,III
Specifications for Product Family

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Loss on drying:
    ≤1.5 % (105°C, 2 h)
  • Titration:
    ≥33.0 % Basic lead (PbO)
  • Chloride (Cl):
    ≤0.003 %
  • Nitrate (NO3):
    ≤0.003 %
  • Calcium (Ca):
    ≤0.01 %
  • Copper (Cu):
    ≤0.002 %
  • Iron (Fe):
    ≤0.002 %
  • Potassium (K):
    ≤0.02 %
  • Sodium (Na):
    ≤0.05 %
  • Insoluble matter:
    ≤0.02 % (in dilute acetic acid)
  • Insoluble matter in water:
    ≤1.0 %
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Plastic bottle
    1 kg
    Log in for availability
    Log in for availability
    Each (1kg)
    Each (1kg) - $74.60
    $74.60
    Each (1kg)
       $74.60

    Supplier Number: AC45322-0010
  • Plastic bottle
    5 kg
    Log in for availability
    Log in for availability
    Each (5kg)
    Each (5kg) - $278.00
    $278.00
    Each (5kg)
       $278.00

    Supplier Number: AC45322-0050

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.