Magnesium hydrogen orthophosphate trihydrate 98%, Extra Pure
TS26478-0010
:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Pack type:Plastic bottle
- Size:1 kg
- Cat. No.:TS26478-0010
- Appearance:White Powder
- Titration Complexometric:≥97.5 %
- Chloride (Cl):≤200 ppm
- Sulfate (SO4):≤300 ppm
- Arsenic (As):≤1 ppm
- Iron (Fe):≤50 ppm
- Heavy metals (as Pb):≤20 ppm
:
:174.33 g/mol
:2.13 g/cm³
:Ambient
:231-823-5
:7782-75-4
:HMgO₄P·3H₂O
:MFCD00150390