Calcium acetate xhydrate 99%, extra pure
Supplier: Thermo Scientific Chemicals
Certificates
About this item
CAS Number:114460-21-8
:5743-26-0
Formula:(H₃CCOO)₂Ca·nH₂O
MDL Number:MFCD00150019
:176.18 g/mol
Storage Temperature:Ambient
EINECS:200-540-9
Specification Test Results
- Appearance:White Powder and/or agglomerates
- Infrared spectrum:Conforms
- Titration Complexometric:98.5 to 101.5 % (on anhydrous substance)
- Water:≤7.0 % (K.F.)
- pH:7.0 to 9.0 (10 % at 20°C)
- Chloride (Cl):≤300 ppm
- Fluoride (F):≤50 ppm
- Sulfate (SO4):≤500 ppm
- Arsenic (As):≤2 ppm
- Iron (Fe):≤20 ppm
- Heavy metals (as Pb):≤10 ppm