Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Magnesium oxide 97+% for analysis

Magnesium oxide 97+% for analysis

Magnesium oxide 97+% for analysis
Certificates
Magnesium oxide 97+% for analysis

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  Magnesia usta
  • CAS Number:1309-48-4
  • Formula:MgO
  • MDL Number:MFCD00011109
  • Molecular Weight:40.3 g/mol
  • Merck Index:13,05700
  • Boiling Pt:3600 °C (760 mmHg)
  • Melting Pt:2800 °C
  • Density:3.58 g/cm³
  • Storage Temperature:Ambient
  • EINECS:215-171-9
Specifications for Product Family

Specification Test Results

  • Appearance:
    White Powder
  • Titration Complexometric:
    ≥97.0 % (on dry substance)
  • Chloride (Cl):
    ≤0.1 %
  • Sulfate (SO4):
    ≤1.0 %
  • Calcium (Ca):
    ≤1.1 %
  • Iron (Fe):
    ≤0.05 %
  • Heavy metals (as Pb):
    ≤0.003 %
  • Soluble matter:
    ≤2 % (in water)
  • Insoluble matter:
    ≤0.1 % (in HCl)
  • Loss on ignition:
    ≤5.0 % (900°C)
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Glass bottle for solids
    250 g
    Log in for availability
    Log in for availability
    Each (250g)
    Each (250g) - $56.40
    $56.40
    Each (250g)
       $56.40

    Supplier Number: AC22253-2500
  • Glass bottle
    1 kg
    Log in for availability
    Log in for availability
    Each (1kg)
    Each (1kg) - $167.00
    $167.00
    Each (1kg)
       $167.00

    Supplier Number: AC22253-0010

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.