Order Entry
Puerto Rico
ContactUsLinkComponent
 
 
di-Ammonium oxalate monohydrate 99.5% for analysis
 
undefined
 
di-Ammonium oxalate monohydrate 99.5% for analysis

 

  • Appearance:
    White Crystalline powder
  • Titration with KMnO4:
    ≥99.4 %
  • Chloride (Cl):
    ≤20 ppm
  • Sulfate (SO4):
    ≤50 ppm
  • Iron (Fe):
    ≤2 ppm
  • Heavy metals (as Pb):
    ≤10 ppm

 

Frequently Bought Together