

Potassium perchlorate 99+% for analysis
Supplier: Thermo Scientific Chemicals
Certificates
About this item
CAS Number:7778-74-7
:
Formula:ClKO₄
MDL Number:MFCD00011362
:138.55 g/mol
Merck Index:13,07738
Melting Pt:400 °C (dec.)
Storage Temperature:Ambient
EINECS:231-912-9
UN:1489
ADR:5.1,II
:GHS07
:GHS03
Specification Test Results
- Appearance:White Crystalline powder
- Titration:≥99.0 %
- Chloride (Cl):≤30 ppm
- Sulfate (SO4):≤10 ppm
- Calcium (Ca):≤50 ppm
- Iron (Fe):≤5 ppm
- Sodium (Na):≤200 ppm
- Heavy metals (as Pb):≤5 ppm
Specification Test Results
Frequently Bought Together