Mercury(II) bromide ACS
Catalog # TS44812-1000
Supplier: Thermo Scientific Chemicals
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle
- Size:100 g
- Cat. No.:TS44812-1000
Specifications
Specification Test Results
- Appearance:White to very light yellow Powder
- Titration Complexometric:≥98.0 %
- Chloride (Cl):≤0.25 %
- Insoluble matter:≤0.05 % (In methanol)
- Residue after reduction:≤0.02 %
Specification Test Results
About this item
:
:360.4 g/mol
Boiling Pt:322 °C (760 mmHg)
Melting Pt:236 °C
Storage Temperature:Ambient
UN:1634
ADR:6.1,II
:GHS06
:GHS08
:GHS09
CAS Number:7789-47-1
Formula:Br₂Hg
MDL Number:MFCD00011040