Potassium dihydrogen phosphate for HPLC
Supplier: Thermo Scientific Chemicals
Certificates
About this item
CAS Number:7778-77-0
:
Formula:KH₂PO₄
MDL Number:MFCD00011401
:136.09 g/mol
Melting Pt:253 °C
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Crystalline powder
- Solubility:(0.1 g/ml) Clear colorless
- UV:(0.5 M in water)
- Titration with NaOH:≥99.4 %
- Gradient analysis:passes test