Antimony(III) fluoride 98%
Supplier: Thermo Scientific Chemicals
About this item
Synonyms: Antimony trifluoride
- CAS Number:7783-56-4
- Formula:F₃Sb
- MDL Number:MFCD00011218
- Molecular Weight:178.76 g/mol
- Merck Index:13,00715
- Boiling Pt:376 °C (1013 hPa)
- Melting Pt:292 °C
- Storage Temperature:Ambient
- EINECS:232-009-2
- UN:2923
- ADR:8,II
2 Options Available Below
Product Details & Documents
Synonyms: Antimony trifluoride
- CAS Number:7783-56-4
- Formula:F₃Sb
- MDL Number:MFCD00011218
- Molecular Weight:178.76 g/mol
- Merck Index:13,00715
- Boiling Pt:376 °C (1013 hPa)
- Melting Pt:292 °C
- Storage Temperature:Ambient
- EINECS:232-009-2
- UN:2923
- ADR:8,II
Specifications for Product Family
Specification Test Results
- Appearance:White to gray Crystalline powder
- Titration Iodimetric:≥97.5 %
Product Family Options
Product Information
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



