Sodium acetate trihydrate for HPLC
Supplier: Thermo Scientific Chemicals
Certificates
About this item
CAS Number:6131-90-4
:
Formula:H₃CCOONa·3H₂O
MDL Number:MFCD00071557
:136.08 g/mol
Melting Pt:58 °C
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Crystals
- UV:(1 M in water)
- pH:7.5 to 9.2
- Titration with HClO4:≥98.5 %
- Insoluble matter:≤0.0005 %