

Sodium dodecyl sulfate (SDS), granular, pure, meets analytical specification of Ph.Eur.
Supplier: Thermo Scientific Chemicals
Certificates
About this item
CAS Number:151-21-3
:
Formula:CH₃(CH₂)₁₁OSO₃Na
MDL Number:MFCD00036175
:288.38 g/mol
Melting Pt:206 °C
Flash Pt:>150 °C
:GHS05
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to light yellow Powder or crystals
- Identification A:Passes test
- Identification B:Passes test
- Identification C:Passes test
- Identification D:Passes test
- Alkalinity:Passes test
- Titration:≥85 %
- NaCl and Na2SO4:≤8 %
- Non-esterified alcohols:≤4 %
Specification Test Results
Frequently Bought Together