
Barium acetate 99+% for analysis
Supplier: Thermo Scientific Chemicals
Certificates
About this item
CAS Number:543-80-6
:
Formula:(H₃CCOO)₂Ba
MDL Number:MFCD00012447
:255.42 g/mol
Merck Index:13,00968
Melting Pt:∼450 °C
Density:2.46 g/cm³
Storage Temperature:Ambient
EINECS:208-849-0
UN:1564
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:White Crystalline powder
- Titration Complexometric:99.0 to 102.0 %
- pH:6.5 to 8.5 (5 % at 20°C)
- Chloride (Cl):≤50 ppm
- Nitrate (NO3):≤20 ppm
- Calcium (Ca):≤50 ppm
- Iron (Fe):≤50 ppm
- Magnesium (Mg):≤10 ppm
- Potassium (K):≤30 ppm
- Sodium (Na):≤50 ppm
- Strontium (Sr):≤0.2 %
- Heavy metals (as Pb):≤5 ppm
Specification Test Results
Frequently Bought Together