Slide jars are ideal for the preservation of pap smear slides, fine needle aspiration (FNA) biopsies, and other small samples.
- Both jars are sold with caps
- Different cap and jar colors
- Sterile and alcohol filled options
Color-coded leakproof screwcaps are vacuum-chamber tested to ensure that our double-sealing screwcaps close tightly every time.