Brady® Aggressive Adhesive Multi-Purpose Nylon Labels with Ribbon, Brady
These pre-sized, high-adhesion nylon cloth labels (B-499) are highly pliable and ideal for tightly curved surfaces such as vials, wires, and cables.
- Permanent acrylic adhesive
- Matte finish
- Autoclavable cryogenic storage labels
Matte white polyamide-coated nylon cloth material has a permanent acrylic adhesive and is thermal transfer printable. Recommended for indoor use. Service temperature of −40 to +193° F (−40 to +90° C).
: UL approved; CSA accepted.