Anti-SARS-CoV-2 Nucleocapsid Mouse Monoclonal Antibody [clone:A20087H](PURE)
Supplier: Biolegend
Certificates
About this item
Specifications
- Catalog No:
- 76508-866
- 76508-864
- Antigen Symbol:
- SARS-CoV-2 Nucleocapsid
- SARS-CoV-2 Nucleocapsid
- Antigen Synonyms:
- Nucleoprotein|Nucleocapsid phosphoprotein|N
- Nucleoprotein|Nucleocapsid phosphoprotein|N
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PURE
- PURE
- Clone:
- A20087H
- A20087H
- Host:
- Mouse
- Mouse
- Isotype:
- IgG2b kappa
- IgG2b kappa
- Immunogen:
- SARS-CoV-2 Nucleocapsid recombinant protein
- SARS-CoV-2 Nucleocapsid recombinant protein
- Purification:
- The antibody was purified by affinity chromatography.
- The antibody was purified by affinity chromatography.
- Concentration:
- 0.5 mg/ml
- 0.5 mg/ml
- Storage Temperature:
- 2...8 °C
- 2...8 °C
Specifications
Frequently Bought Together