Anti-SARS-CoV-2 S Protein RBD Mouse Monoclonal Antibody [clone:A20100I](ULEAF)
Supplier: Biolegend
Certificates
About this item
Specifications
- Catalog No:
- 76508-610
- 76508-608
- Antigen Symbol:
- SARS-CoV-2 S Protein RBD
- SARS-CoV-2 S Protein RBD
- Antigen Synonyms:
- S1 RBD|Spike Protein RBD|S1 Receptor-binding Domain|Spike Receptor-binding Domain
- S1 RBD|Spike Protein RBD|S1 Receptor-binding Domain|Spike Receptor-binding Domain
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Ultra-LEAF™
- Ultra-LEAF™
- Clone:
- A20100I
- A20100I
- Host:
- Mouse
- Mouse
- Isotype:
- IgG2a kappa
- IgG2a kappa
- Immunogen:
- SARS-CoV-2
- SARS-CoV-2
- Storage Temperature:
- 2...8 °C
- 2...8 °C