Anti-NRF2 Rat Monoclonal Antibody [clone:W19086B](PURE)
Supplier: Biolegend
Certificates
About this item
Specifications
- Catalog No:
- 76508-414
- 76508-416
- Antigen Symbol:
- NRF2
- NRF2
- Antigen Synonyms:
- HEBP1|IMDDHH|Erythroid 2 Like 2|NF-E2-Related Factor 2
- HEBP1|IMDDHH|Erythroid 2 Like 2|NF-E2-Related Factor 2
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PURE
- PURE
- Clone:
- W19086B
- W19086B
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rat
- Rat
- Isotype:
- IgG2a kappa
- IgG2a kappa
- Immunogen:
- Partial recombinant human NRF2 protein
- Partial recombinant human NRF2 protein
- Purification:
- The antibody was purified by affinity chromatography.
- The antibody was purified by affinity chromatography.
- Concentration:
- 0.5 mg/ml
- 0.5 mg/ml
- Storage Temperature:
- 2...8 °C
- 2...8 °C
Specifications
Frequently Bought Together