Anti-SARS-CoV-2 Spike S1 Human Monoclonal Antibody [clone: HC2001]
Supplier: Genscript
Certificates
About this item
The product is specific for SARS-CoV-2 Spike Protein S1 subunit and its RBD domain.
- COVID-19 antibodies
- Validated for ELISA, WB, FACS and SARS-CoV-2 virus neutralization assay
- Control antibodies and various antibody pairs available for ELISA-based detection
- Neutralizing antibody standard to calibrate quantitative assay of SARS-CoV-2 neutralizing antibodies in samples
Genscript offers site-specific neutralizing antibodies, neutralizing antibody standards and control antibodies.
Type: Primary
Antigen: SARS-CoV-2 Spike S1
Clonality: Monoclonal
Clone: HC2001
Conjugation: Unconjugated
Public Immunogen Range:
Host: Human
Isotype: IgG1
Reactivity: Human
Specifications
- Catalog No:
- 76510-092
- 76510-094
- Antigen Symbol:
- SARS-CoV-2 Spike S1
- SARS-CoV-2 Spike S1
- Antigen Name:
- SARS-CoV-2 Spike S1
- SARS-CoV-2 Spike S1
- Antigen Synonyms:
- Receptor-binding domain/RBD
- Receptor-binding domain/RBD
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- HC2001
- HC2001
- Host:
- Human
- Human
- Gene ID:
- MN908947
- MN908947
- Isotype:
- IgG1
- IgG1
- ELISA:
- Yes
- Yes
- Antibody Fragment:
- Whole Molecule
- Whole Molecule
- Modification:
- Humanization
- Humanization
- Epitope:
- Confidentiality
- Confidentiality
- Immunogen:
- Recombinant SARS-CoV-2 RBD Protein
- Recombinant SARS-CoV-2 RBD Protein
- Amino Acid Number:
- Confidentiality
- Confidentiality
- Sequence:
- Confidentiality
- Confidentiality
- Format:
- Purified antibody
- Purified antibody
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 150 kDa
- 150 kDa
- Purification:
- Protein A affinity column
- Protein A affinity column
- Concentration:
- 1 mg/ml
- 1 mg/ml
- Cross Adsorption:
- No
- No
- Storage Buffer:
- PBS, pH 7.4, containing 0.02% sodium azide
- PBS, pH 7.4, containing 0.02% sodium azide
- Storage Temperature:
- Store at -20 °C. This product is stable for 1 year upon receipt, when handled and stored as instructed. Avoid repeated freezing and thawing cycles.
- Store at -20 °C. This product is stable for 1 year upon receipt, when handled and stored as instructed. Avoid repeated freezing and thawing cycles.
- Shipping Temperature:
- Ice
- Ice