Anti-ABCC11 Rabbit Polyclonal Antibody
Produced in rabbits immunized with E. coli-derived Human ABCC11 fragment, and purified by antigen affinity chromatography.
- Catalog No:
- 76535-338
- Antigen Symbol:
- ABCC11
- Antigen Name:
- ATP binding cassette subfamily C member 11
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human
- Host:
- Rabbit
- Isotype:
- IgG
- ImmunoChemistry:
- Yes
- Form:
- Liquid
- Storage Buffer:
- PBS, pH 7.0 with 0.03% Proclin300
- Shipping Temperature:
- Ambient