Anti-ACRV1 Rabbit Polyclonal Antibody
Supplier: Sino Biological
Certificates
About this item
Produced in rabbits immunized with purified, recombinant Human SP-10/ACRV1 (rh SP-10/ACRV1; Catalog#11789-H08H; P26436-1; Gln22-Ile265). SP-10/ACRV1 specific IgG was purified by Human SP-10/ACRV1 affinity chromatography.
Specifications
- Catalog No:
- 76535-588
- Antigen Symbol:
- ACRV1
- Antigen Name:
- Acrosomal vesicle protein 1
- Antigen Synonyms:
- D11S4365|SP-10|SPACA2
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human
- Host:
- Rabbit
- Isotype:
- IgG
- ELISA:
- Yes
- ImmunoChemistry:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm Filtered solution in PBS
- Shipping Temperature:
- Ambient