Anti-CD226 (DNAM-1) Mouse Monoclonal Antibody [clone: 11A8] (Brilliant Violet® 605)
76522-050
- Antigen Symbol:CD226 (DNAM-1)
- Clonality:Monoclonal
- Clone:11A8
- Conjugation:Brilliant Violet® 605
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Baboon,African green monkey,Cynomolgus monkey,Rhesus monkey,Human
- Size:100 tests
- Antigen Synonyms:TLiSA1|PTA1|DNAM-1
- Storage Buffer:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Storage Temperature:2...8 °C
- Tested Applications:FC
- Purification:The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 605™ under optimal conditions.
- Cat. No.:76522-050