Anti-HLA-DR Mouse Monoclonal Antibody [clone:L243](KIRAVIA Blue 520™)
Supplier: Biolegend
Certificates
About this item
Specifications
- Catalog No:
- 76508-882
- 76508-880
- Antigen Symbol:
- HLA-DR
- HLA-DR
- Antigen Synonyms:
- Major Histocompatibility Class II|MHC class II
- Major Histocompatibility Class II|MHC class II
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- KIRAVIA Blue 520™
- KIRAVIA Blue 520™
- Clone:
- L243
- L243
- Reactivity:
- Marmoset, Baboon, Tamarin, African green monkey, Cynomolgus monkey, Rhesus monkey, Chimpanzee, Human, Dog, Squirrel monkey
- Marmoset, Baboon, Tamarin, African green monkey, Cynomolgus monkey, Rhesus monkey, Chimpanzee, Human, Dog, Squirrel monkey
- Host:
- Mouse
- Mouse
- Isotype:
- IgG2a kappa
- IgG2a kappa
- Purification:
- The antibody was purified by affinity chromatography and conjugated with KIRAVIA Blue 520™ under optimal conditions.
- The antibody was purified by affinity chromatography and conjugated with KIRAVIA Blue 520™ under optimal conditions.
- Storage Temperature:
- 2...8 °C
- 2...8 °C