Anti-Ap2a2 Rabbit Polyclonal Antibody
Supplier: Sino Biological
Certificates
About this item
Produced in rabbits immunized with E. coli-derived Human AP2A2 fragment, and purified by antigen affinity chromatography.
Specifications
- Catalog No:
- 76536-748
- Antigen Symbol:
- Ap2a2
- Antigen Name:
- adaptor related protein complex 2 subunit alpha 2
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human
- Host:
- Rabbit
- Isotype:
- IgG
- Western Blot:
- Yes
- ImmunoChemistry:
- Yes
- ImmunoPrecipitation:
- Yes
- Form:
- Liquid
- Storage Buffer:
- PBS, pH 7.0 with 0.03% Proclin300
- Shipping Temperature:
- Ambient