Anti-MERTK (Mer) Rat Monoclonal Antibody [clone:2B10C42](KIRAVIA Blue 520™)
- Catalog No:
- 76508-636
- 76508-638
- Antigen Symbol:
- MERTK (Mer)
- MERTK (Mer)
- Antigen Synonyms:
- Mer|Nyk|Eyk|nmf12
- Mer|Nyk|Eyk|nmf12
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- KIRAVIA Blue 520™
- KIRAVIA Blue 520™
- Clone:
- 2B10C42
- 2B10C42
- Reactivity:
- Mouse
- Mouse
- Host:
- Rat
- Rat
- Isotype:
- IgG2a kappa
- IgG2a kappa
- Immunogen:
- Mouse MERTK extracellular domain
- Mouse MERTK extracellular domain
- Purification:
- The antibody was purified by affinity chromatography and conjugated with KIRAVIA Blue 520™ under optimal conditions.
- The antibody was purified by affinity chromatography and conjugated with KIRAVIA Blue 520™ under optimal conditions.
- Concentration:
- 0.2 mg/ml
- 0.2 mg/ml
- Storage Temperature:
- 2...8 °C
- 2...8 °C