Anti-CD45 Rat Monoclonal Antibody [clone:S18009D](Alexa Fluor® 594)
- Catalog No:
- 76508-552
- 76508-548
- Antigen Symbol:
- CD45
- CD45
- Antigen Synonyms:
- Ly-5|T200|LCA
- Ly-5|T200|LCA
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Alexa Fluor® 594
- Alexa Fluor® 594
- Clone:
- S18009D
- S18009D
- Reactivity:
- Mouse
- Mouse
- Host:
- Rat
- Rat
- Isotype:
- IgG2a kappa
- IgG2a kappa
- Immunogen:
- Recombinant mouse CD45
- Recombinant mouse CD45
- Purification:
- The antibody was purified by affinity chromatography and conjugated with Alexa Fluor® 594 under optimal conditions.
- The antibody was purified by affinity chromatography and conjugated with Alexa Fluor® 594 under optimal conditions.
- Concentration:
- 0.5 mg/ml
- 0.5 mg/ml
- Storage Temperature:
- 2...8 °C
- 2...8 °C