ContactUsLinkComponent
Decontamination bags, activated charcoal, Biotium
Decontamination bags, activated charcoal, Biotium
Catalog # 89427-060
Supplier:  Biotium
CAS Number:  
undefined
Decontamination bags, activated charcoal, Biotium
Catalog # 89427-060
Supplier:  Biotium
Supplier Number:  22007
CAS Number:  
Specifications
  • Description:
    Activated Charcoal Decontamination Bags
  • Size:
    25 bags
  • Cat. No.:
    89427-060
  • Supplier no.:
    22007
Specifications
About this item
Activated charcoal retains ethidium bromide, GelRed™ and GelGreen™ efficiently in a convenient bag for decontamination of used gel staining solutions. Each bag has a total capacity of at least 10 mg ethidium bromide.
  • Remove mutagenic dyes (ie, ethidium bromide) from gel staining solutions
  • Use to clean up any dyes from a solution to allow drain disposal
  • No-mess, convenient bag
  • Reusable bag capacity of at least 10 mg EtBr