Order Entry
Puerto Rico
ContactUsLinkComponent
 
Mag-Bind® FFPE DNA/RNA 96 Kit
 
undefined
Mag-Bind® FFPE DNA/RNA 96 Kit

 

  • Method / Format:
    Magnetic beads
  • Prep size:
    Scalable
  • Refine target molecule:
    Total RNA
  • Sample type:
    FFPE tissues
  • Target molecule:
    RNA

 

Frequently Bought Together